missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%, Solstice

Catalog No. 6000534
Change view
Click to view available options
Quantity:
19 L
Packaging:
Metal Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000534 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000534 Supplier Solstice Supplier No. PP10819

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

CAS Max % 100.0000%, ≤100.0000%
Density 0.713 g/cm3
Quantity 19 L
Flash Point −45°C
Packaging Metal Can
UN Number UN1155
Water 0.03% max.
Beilstein 1696894
Boiling Point 35°C
Solubility Information 689% in Water at 20°C
Formula Weight 74.12
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Ethyl Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether