missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ether (Laboratory), Fisher Chemical™

Catalog No. p-215990
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Aluminum Can
Steel Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E1341 1 L Aluminum Can
E1344 4 L Aluminum Can
E13420 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E1341 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. FLE1341
Only null left

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Density 0.714g/cm³
Quantity 1 L
Flash Point -45°C (-111°F)
Identification Pass Test
Packaging Aluminum Can
Physical Form Liquid
Vapor Pressure 587mbar at 20°C
Linear Formula C2H5OC2H5
Viscosity 0.2448 cP at 20°C
Boiling Point 34.6°C
Formula Weight 74.12
Grade Laboratory
Chemical Name or Material Ethyl Ether
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Handle under an inert atmosphere. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:4
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid