missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ether (Diethyl Ether), Reagents

Catalog No. p-200021131
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338001 20 L
01338000 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338001 Supplier Labchem Inc Supplier No. C119850020E6
Only null left

Ethyl Ether (Diethyl Ether), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym Ether, Diethyl Ether, Ethoxyethane
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Boiling Point 35°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7134
Grade ACS
Chemical Name or Material Ethyl Ether (Diethyl Ether)