missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Ether (Diethyl Ether), Reagents
Specifications
Specifications
| Melting Point | -116°C |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.7134 |
| Boiling Point | 35°C |
| Grade | ACS |
| Chemical Name or Material | Ethyl Ether (Diethyl Ether) |