missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Ether, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains no preservative, >99.9%, Solstice
Specifications
Specifications
| Absorbance | 0.08 AU max. at 250 nm (UV), 0.01 AU max. at 275 nm (UV), 0.005 AU max. at 300 nm (UV) |
| CAS Max % | 100.0000% |
| Density | 0.713 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | −45°C |
| Packaging | Glass Bottle |
| UN Number | UN1155 |
| Beilstein | 1696894 |
| Boiling Point | 35°C |
| Formula Weight | 74.12 |
| Show More |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether |