missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%

Catalog No. AAH2724406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2724406 5 g
AAH2724414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2724406 Supplier Thermo Scientific Chemicals Supplier No. H2724406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 932750-29-3 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932750-29-3
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD01743485
InChI Key YKXFOGAYPIPTKF-BQYQJAHWSA-N
Synonym ethyl 2e-2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxycrotonate, ethyl 2-cyano-3-ethoxybut-2-enoate, 2-butenoic acid, 2-cyano-3-ethoxy-, ethyl ester, ethyl e-2-cyano-3-ethoxybut-2-enoate, e-ethyl 2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxy-2-butenoate, e-2-cyano-3-ethoxycrotonic acid ethyl ester, e-2-cyano-3-ethoxy-2-butenoic acid ethyl ester
PubChem CID 6387109
IUPAC Name ethyl (E)-2-cyano-3-ethoxybut-2-enoate
SMILES CCOC(=C(C#N)C(=O)OCC)C

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
UN Number UN3439
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material Ethyl (E)-2-cyano-3-ethoxycrotonate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only