missing translation for 'onlineSavingsMsg'
Learn More

Ethyl DL-Mandelate 95.0+%, TCI America™

Catalog No. M0039500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0039500G 500 g
M003925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0039500G Supplier TCI America Supplier No. M0039500G
Only null left

Chemical Identifiers

CAS 774-40-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00004494
InChI Key SAXHIDRUJXPDOD-UHFFFAOYSA-N
Synonym ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
PubChem CID 13050
ChEBI CHEBI:38750
IUPAC Name ethyl 2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Color White-Yellow
Boiling Point 133°C
Quantity 500 g
Formula Weight 180.20
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl DL-Mandelate
EINECSNumber (3)-3841
RTECSNumber OO7397000
TSCA No