missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl DL-Mandelate 95.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 133°C |
| Quantity | 25 g |
| Formula Weight | 180.20 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Ethyl DL-Mandelate |
Safety and Handling
| EINECSNumber | (3)-3841 |
| RTECSNumber | OO7397000 |
| TSCA | No |