missing translation for 'onlineSavingsMsg'
Learn More

Ethyl DL-Leucate 98.0+%, TCI America™

Catalog No. p-7129159
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08025G 5 g
E080225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08025G Supplier TCI America Supplier No. E08025G

Chemical Identifiers

CAS 10348-47-7
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
InChI Key QRHOWVDPHIXNEN-UHFFFAOYSA-N
Synonym DL-Leucic Acid Ethyl Ester, Ethyl DL-2-Hydroxy-4-methylvalerate, DL-2-Hydroxy-4-methylvaleric Acid Ethyl Ester
PubChem CID 112030
IUPAC Name ethyl 2-hydroxy-4-methylpentanoate
SMILES CCOC(=O)C(CC(C)C)O

Specifications

Color Colorless
Boiling Point 185°C
Quantity 5 g
Formula Weight 160.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl DL-Leucate
EINECSNumber (2)-1430
TSCA No
Recommended Storage Refrigerator