missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl DL-3-Acetoxybutyrate 98.0+%, TCI America™
$54.06 - $225.56
Chemical Identifiers
| CAS | 27846-49-7 |
|---|---|
| Molecular Formula | C8H14O4 |
| Molecular Weight (g/mol) | 174.20 |
| MDL Number | MFCD00059378 |
| InChI Key | JFWYJXYRFBRWSH-UHFFFAOYNA-N |
| Synonym | DL-3-Acetoxybutyric Acid Ethyl Ester |
| PubChem CID | 537574 |
| IUPAC Name | ethyl 3-(acetyloxy)butanoate |
| SMILES | CCOC(=O)CC(C)OC(C)=O |
Chemical Identifiers
| 27846-49-7 | |
| 174.20 | |
| JFWYJXYRFBRWSH-UHFFFAOYNA-N | |
| 537574 | |
| CCOC(=O)CC(C)OC(C)=O |
| C8H14O4 | |
| MFCD00059378 | |
| DL-3-Acetoxybutyric Acid Ethyl Ester | |
| ethyl 3-(acetyloxy)butanoate |
Specifications
| 27846-49-7 | |
| 94°C | |
| MFCD00059378 | |
| 3272 | |
| JFWYJXYRFBRWSH-UHFFFAOYNA-N | |
| ethyl 3-(acetyloxy)butanoate | |
| 537574 | |
| ≥98.0% (GC) | |
| Ethyl DL-3-Acetoxybutyrate |
| Yellow | |
| C8H14O4 | |
| 5 mL | |
| DL-3-Acetoxybutyric Acid Ethyl Ester | |
| CCOC(=O)CC(C)OC(C)=O | |
| 174.20 | |
| 174.20 | |
| Liquid |
Safety and Handling
TSCA : No