missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Dimethylphosphonoacetate 97.0+%, TCI America™
Supplier: TCI America E116025G
Specifications
| Ethyl Dimethylphosphonoacetate | |
| Colorless | |
| C6H13O5P | |
| 25 g | |
| HUNISAHOCCASGM-UHFFFAOYSA-N | |
| ethyl 2-dimethoxyphosphorylacetate | |
| 67555 | |
| ≥97.0% (GC) |
| 311-46-6 | |
| 135°C | |
| MFCD00015677 | |
| Dimethylphosphonoacetic Acid Ethyl Ester | |
| CCOC(=O)CP(=O)(OC)OC | |
| 196.139 | |
| 196.14 | |
| Liquid |
Chemical Identifiers
| 311-46-6 | |
| 196.139 | |
| HUNISAHOCCASGM-UHFFFAOYSA-N | |
| 67555 | |
| CCOC(=O)CP(=O)(OC)OC |
| C6H13O5P | |
| MFCD00015677 | |
| Dimethylphosphonoacetic Acid Ethyl Ester | |
| ethyl 2-dimethoxyphosphorylacetate |
Safety and Handling
TSCA : No