missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Diformylacetate 97.0+%, TCI America™
$72.70 - $294.52
Chemical Identifiers
| CAS | 80370-42-9 |
|---|---|
| Molecular Formula | C6H8O4 |
| Molecular Weight (g/mol) | 144.126 |
| MDL Number | MFCD11112084 |
| InChI Key | HMFLBGNCDZYITR-UHFFFAOYSA-N |
| Synonym | Diformylacetic Acid Ethyl Ester, Ethyl 2-Formyl-3-oxopropionate, 2-Formyl-3-oxopropionic Acid Ethyl Ester |
| PubChem CID | 11029955 |
| IUPAC Name | ethyl 2-formyl-3-oxopropanoate |
| SMILES | CCOC(=O)C(C=O)C=O |
Chemical Identifiers
| 80370-42-9 | |
| 144.126 | |
| HMFLBGNCDZYITR-UHFFFAOYSA-N | |
| 11029955 | |
| CCOC(=O)C(C=O)C=O |
| C6H8O4 | |
| MFCD11112084 | |
| Diformylacetic Acid Ethyl Ester, Ethyl 2-Formyl-3-oxopropionate, 2-Formyl-3-oxopropionic Acid Ethyl Ester | |
| ethyl 2-formyl-3-oxopropanoate |
Specifications
| 80370-42-9 | |
| 32°C | |
| MFCD11112084 | |
| Diformylacetic Acid Ethyl Ester, Ethyl 2-Formyl-3-oxopropionate, 2-Formyl-3-oxopropionic Acid Ethyl Ester | |
| CCOC(=O)C(C=O)C=O | |
| 144.126 | |
| 144.13 | |
| Liquid |
| Yellow | |
| C6H8O4 | |
| 1 g | |
| HMFLBGNCDZYITR-UHFFFAOYSA-N | |
| ethyl 2-formyl-3-oxopropanoate | |
| 11029955 | |
| ≥97.0% (T) | |
| Ethyl Diformylacetate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator