missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Difluoroacetate 98.0+%, TCI America™

Catalog No. p-7127337
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24985G 5 g
D249825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24985G Supplier TCI America Supplier No. D24985G
Only null left

Chemical Identifiers

CAS 454-31-9
Molecular Formula C4H6F2O2
Molecular Weight (g/mol) 124.087
MDL Number MFCD00013578
InChI Key GZKHDVAKKLTJPO-UHFFFAOYSA-N
Synonym ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat
PubChem CID 9961
IUPAC Name ethyl 2,2-difluoroacetate
SMILES CCOC(=O)C(F)F

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 g
UN Number 2924
Formula Weight 124.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Difluoroacetate
EINECSNumber (2)-3848
TSCA Yes