missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Dibromofluoroacetate 97.0+%, TCI America™

Catalog No. p-7127352
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25305G 5 g
D253025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25305G Supplier TCI America Supplier No. D25305G
Only null left

Chemical Identifiers

CAS 565-53-7
Molecular Formula C4H5Br2FO2
Molecular Weight (g/mol) 263.89
MDL Number MFCD00042068
InChI Key COYRDMUQLSIAEX-UHFFFAOYSA-N
Synonym ethyl dibromofluoroacetate, dibromofluoroacetic acid ethyl ester, ethyldibromofluoroacetate, 2,2-dibromo-1-ethoxy-2-fluoroethan-1-one, acmc-1aqw2, ethyl 2,2-dibromo-2-fluoracetate, ethyl 2,2-dibromo-2-fluoro-acetate
PubChem CID 2737168
IUPAC Name ethyl 2,2-dibromo-2-fluoroacetate
SMILES CCOC(=O)C(F)(Br)Br

Specifications

Color Yellow
Boiling Point 173°C
Quantity 5 g
UN Number 2927
Formula Weight 263.89
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Dibromofluoroacetate