missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl dibromofluoroacetate, 97%
Description
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.
Solubility
Miscible with most organic solvents. Immiscible with water.
Notes
Incompatible with oxidizing agents and bases.
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.
Solubility
Miscible with most organic solvents. Immiscible with water.
Notes
Incompatible with oxidizing agents and bases.
Specifications
Specifications
| Density | 1.894 |
| Boiling Point | 82°C (33 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.4635 |
| Linear Formula | Br2CFCO2CH2CH3 |
| Quantity | 5 g |
| UN Number | UN2922 |
| Beilstein | 1762048 |
| Solubility Information | Miscible with most organic solvents. Immiscible with water. |
| Formula Weight | 263.89 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only