missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Diacetoacetate 96.0+%, TCI America™

Catalog No. D21705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21705G 5 g
D217025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21705G Supplier TCI America Supplier No. D21705G

Chemical Identifiers

CAS 603-69-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009864
InChI Key YMCDYRGMTRCAPZ-UHFFFAOYSA-N
PubChem CID 79063
IUPAC Name ethyl 2-acetyl-3-oxobutanoate
SMILES CCOC(=O)C(C(=O)C)C(=O)C

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
Synonym ethyl diacetoacetate, ethyl 2-acetylacetoacetate, butanoic acid, 2-acetyl-3-oxo-, ethyl ester, 3-ethoxycarbonylacetylacetone, ethyl diacetylacetate, ethyl diacetyl acetate, ethyl diaceto acetate, 2-acetyl-3-oxo-butyric acid ethyl ester, butanoic acid,2-acetyl-3-oxo-, ethyl ester, ethyldiacetoacetate
Formula Weight 172.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Diacetoacetate