missing translation for 'onlineSavingsMsg'
Learn More

Ethyl diacetoacetate, 97%

CAS: 603-69-0 | C8H12O4 | 172.18 g/mol

$109.81 - $353.05

Chemical Identifiers

CAS 603-69-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009864
InChI Key YMCDYRGMTRCAPZ-UHFFFAOYSA-N
PubChem CID 79063
IUPAC Name ethyl 2-acetyl-3-oxobutanoate
SMILES CCOC(=O)C(C(=O)C)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2014909
View Documents
Thermo Scientific Chemicals
B2014909
10 g
Each for $109.81
Only null left
 
AAB2014918
View Documents
Thermo Scientific Chemicals
B2014918
50 g
Each for $353.05
Only null left
 
Description

Description

Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

603-69-0
172.18
YMCDYRGMTRCAPZ-UHFFFAOYSA-N
ethyl 2-acetyl-3-oxobutanoate
C8H12O4
MFCD00009864
79063
CCOC(=O)C(C(=O)C)C(=O)C

Specifications

603-69-0
104°C to 105°C (17 mmHg)
C8H12O4
MFCD00009864
775014
Not miscible in water.
CCOC(=O)C(C(=O)C)C(=O)C
172.18
172.18
Ethyl diacetoacetate
1.104
85°C (185°F)
1.472
10 g
ethyl diacetoacetate, ethyl 2-acetylacetoacetate, butanoic acid, 2-acetyl-3-oxo-, ethyl ester, 3-ethoxycarbonylacetylacetone, ethyl diacetylacetate, ethyl diacetyl acetate, ethyl diaceto acetate, 2-acetyl-3-oxo-butyric acid ethyl ester, butanoic acid,2-acetyl-3-oxo-, ethyl ester, ethyldiacetoacetate
YMCDYRGMTRCAPZ-UHFFFAOYSA-N
ethyl 2-acetyl-3-oxobutanoate
79063
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 210-053-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only