missing translation for 'onlineSavingsMsg'
Learn More

Ethyl diacetoacetate, 97%

Catalog No. AAB2014918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2014918 50 g
AAB2014909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2014918 Supplier Thermo Scientific Chemicals Supplier No. B2014918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 603-69-0 | C8H12O4 | 172.18 g/mol

Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 603-69-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009864
InChI Key YMCDYRGMTRCAPZ-UHFFFAOYSA-N
PubChem CID 79063
IUPAC Name ethyl 2-acetyl-3-oxobutanoate
SMILES CCOC(=O)C(C(=O)C)C(=O)C

Specifications

Density 1.104
Boiling Point 104°C to 105°C (17 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.472
Quantity 50 g
Beilstein 775014
Synonym ethyl diacetoacetate, ethyl 2-acetylacetoacetate, butanoic acid, 2-acetyl-3-oxo-, ethyl ester, 3-ethoxycarbonylacetylacetone, ethyl diacetylacetate, ethyl diacetyl acetate, ethyl diaceto acetate, 2-acetyl-3-oxo-butyric acid ethyl ester, butanoic acid,2-acetyl-3-oxo-, ethyl ester, ethyldiacetoacetate
Solubility Information Not miscible in water.
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Ethyl diacetoacetate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-053-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only