missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl diacetoacetate, 97%
Description
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
| Density | 1.104 |
| Boiling Point | 104°C to 105°C (17 mmHg) |
| Flash Point | 85°C (185°F) |
| Refractive Index | 1.472 |
| Quantity | 10 g |
| Beilstein | 775014 |
| Synonym | ethyl diacetoacetate, ethyl 2-acetylacetoacetate, butanoic acid, 2-acetyl-3-oxo-, ethyl ester, 3-ethoxycarbonylacetylacetone, ethyl diacetylacetate, ethyl diacetyl acetate, ethyl diaceto acetate, 2-acetyl-3-oxo-butyric acid ethyl ester, butanoic acid,2-acetyl-3-oxo-, ethyl ester, ethyldiacetoacetate |
| Solubility Information | Not miscible in water. |
| Formula Weight | 172.18 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H227-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H227 Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
| missing translation for 'healthHazard3' | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 210-053-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only