missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Decanoate 98.0+%, TCI America™
$55.92 - $143.54
Chemical Identifiers
| CAS | 110-38-3 |
|---|---|
| Molecular Formula | C12H24O2 |
| Molecular Weight (g/mol) | 200.322 |
| MDL Number | MFCD00009581 |
| InChI Key | RGXWDWUGBIJHDO-UHFFFAOYSA-N |
| Synonym | ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo |
| PubChem CID | 8048 |
| ChEBI | CHEBI:87430 |
| IUPAC Name | ethyl decanoate |
| SMILES | CCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 110-38-3 | |
| 200.322 | |
| RGXWDWUGBIJHDO-UHFFFAOYSA-N | |
| 8048 | |
| ethyl decanoate |
| C12H24O2 | |
| MFCD00009581 | |
| ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo | |
| CHEBI:87430 | |
| CCCCCCCCCC(=O)OCC |
Specifications
| 110-38-3 | |
| Yellow | |
| C12H24O2 | |
| 25 mL | |
| RGXWDWUGBIJHDO-UHFFFAOYSA-N | |
| ethyl decanoate | |
| 8048 | |
| 200.32 | |
| Liquid |
| -20°C | |
| 243°C | |
| MFCD00009581 | |
| ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo | |
| CCCCCCCCCC(=O)OCC | |
| 200.322 | |
| CHEBI:87430 | |
| ≥98.0% (GC) | |
| Ethyl Decanoate |
Safety and Handling
EINECSNumber : (2)-0782&(2)-0798
RTECSNumber : HD9420000
TSCA : Yes