missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Decanoate 98.0+%, TCI America™

Catalog No. D002225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D002225ML 25 mL
D0022500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D002225ML Supplier TCI America Supplier No. D002225ML

Chemical Identifiers

CAS 110-38-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00009581
InChI Key RGXWDWUGBIJHDO-UHFFFAOYSA-N
Synonym ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
PubChem CID 8048
ChEBI CHEBI:87430
IUPAC Name ethyl decanoate
SMILES CCCCCCCCCC(=O)OCC

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 243°C
Quantity 25 mL
Formula Weight 200.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Decanoate
EINECSNumber (2)-0782&(2)-0798
RTECSNumber HD9420000
TSCA Yes