missing translation for 'onlineSavingsMsg'
Learn More

Ethyl decanoate, 99%

CAS: 110-38-3 | C12H24O2 | 200.322 g/mol

$60.38 - $804.97

Chemical Identifiers

CAS 110-38-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00009581
InChI Key RGXWDWUGBIJHDO-UHFFFAOYSA-N
Synonym ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
PubChem CID 8048
ChEBI CHEBI:87430
IUPAC Name ethyl decanoate
SMILES CCCCCCCCCC(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1065722
View Documents
Thermo Scientific Chemicals
A1065722
100 g
Each for $60.38
Only null left
 
AAA1065736
View Documents
Thermo Scientific Chemicals
A1065736
500 g
Each for $194.18
Only null left
 
AAA106570E
View Documents
Thermo Scientific Chemicals
A106570E
2500 g
Each for $804.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-38-3
200.322
RGXWDWUGBIJHDO-UHFFFAOYSA-N
8048
ethyl decanoate
C12H24O2
MFCD00009581
ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
CHEBI:87430
CCCCCCCCCC(=O)OCC

Specifications

110-38-3
0.862
102°C (216°F)
1.425
100 g
14,3776
RGXWDWUGBIJHDO-UHFFFAOYSA-N
ethyl decanoate
8048
200.32
Ethyl decanoate
-20°C
250°C to 252°C
C12H24O2
MFCD00009581
1762128
ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
CCCCCCCCCC(=O)OCC
200.322
CHEBI:87430
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-761-9

RTECSNumber : HD9420000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only