missing translation for 'onlineSavingsMsg'
Learn More

Ethyl decanoate, 99%

Catalog No. AAA1065722
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1065722 100 g
AAA1065736 500 g
AAA106570E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1065722 Supplier Thermo Scientific Chemicals Supplier No. A1065722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-38-3 | C12H24O2 | 200.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-38-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00009581
InChI Key RGXWDWUGBIJHDO-UHFFFAOYSA-N
Synonym ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
PubChem CID 8048
ChEBI CHEBI:87430
IUPAC Name ethyl decanoate
SMILES CCCCCCCCCC(=O)OCC

Specifications

Melting Point -20°C
Density 0.862
Boiling Point 250°C to 252°C
Flash Point 102°C (216°F)
Refractive Index 1.425
Quantity 100 g
Beilstein 1762128
Merck Index 14,3776
Formula Weight 200.32
Percent Purity 99%
Chemical Name or Material Ethyl decanoate
Show More Show Less
EINECSNumber 203-761-9
RTECSNumber HD9420000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only