missing translation for 'onlineSavingsMsg'
Learn More

Ethyl D-(-)-Mandelate 98.0+%, TCI America™

Catalog No. M13445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13445G 5 g
M134425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13445G Supplier TCI America Supplier No. M13445G
Only null left

Chemical Identifiers

CAS 10606-72-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00064248
InChI Key SAXHIDRUJXPDOD-SECBINFHSA-N
Synonym D-(-)-Mandelic Acid Ethyl Ester
PubChem CID 6951556
ChEBI CHEBI:78406
IUPAC Name ethyl (2R)-2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl D-(-)-Mandelate
EINECSNumber (3)-3841
TSCA No