missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyanoformate 98.0+%, TCI America™

Catalog No. p-7124883
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12665G 5 g
C126625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12665G Supplier TCI America Supplier No. C12665G
Only null left

Chemical Identifiers

CAS 623-49-4
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001836
InChI Key MSMGXWFHBSCQFB-UHFFFAOYSA-N
Synonym carbonocyanidic acid, ethyl ester, ethyl carbonocyanidate, 2-ethoxy-2-oxoacetonitrile, cyanoformic acid ethyl ester, ethyl carbonocyanidoate, formic acid, cyano-, ethyl ester, cyanocarbonyloxyethane, ethylcyanidocarbonat, ethyl cyanoformate, ethyl cyanocarbonate
PubChem CID 69336
IUPAC Name ethyl cyanoformate
SMILES CCOC(=O)C#N

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
UN Number 3275
Formula Weight 99.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cyanoformate