missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyanoformate, 99%

CAS: 623-49-4 | C4H5NO2 | 99.09 g/mol

$120.99 - $120.99

Chemical Identifiers

CAS 623-49-4
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001836
InChI Key MSMGXWFHBSCQFB-UHFFFAOYSA-N
Synonym carbonocyanidic acid, ethyl ester, ethyl carbonocyanidate, 2-ethoxy-2-oxoacetonitrile, cyanoformic acid ethyl ester, ethyl carbonocyanidoate, formic acid, cyano-, ethyl ester, cyanocarbonyloxyethane, ethylcyanidocarbonat, ethyl cyanoformate, ethyl cyanocarbonate
PubChem CID 69336
IUPAC Name ethyl cyanoformate
SMILES CCOC(=O)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC118300100
SDS View Documents
Thermo Scientific Chemicals
118300100
10 g Glass bottle
Each for $120.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-49-4
99.09
MSMGXWFHBSCQFB-UHFFFAOYSA-N
69336
CCOC(=O)C#N
C4H5NO2
MFCD00001836
carbonocyanidic acid, ethyl ester, ethyl carbonocyanidate, 2-ethoxy-2-oxoacetonitrile, cyanoformic acid ethyl ester, ethyl carbonocyanidoate, formic acid, cyano-, ethyl ester, cyanocarbonyloxyethane, ethylcyanidocarbonat, ethyl cyanoformate, ethyl cyanocarbonate
ethyl cyanoformate

Specifications

623-49-4
1.0000g/mL
24°C
98.5% min. (GC)
C4H5NO2
NCCO2C2H5
10 g
07,145
carbonocyanidic acid, ethyl ester, ethyl carbonocyanidate, 2-ethoxy-2-oxoacetonitrile, cyanoformic acid ethyl ester, ethyl carbonocyanidoate, formic acid, cyano-, ethyl ester, cyanocarbonyloxyethane, ethylcyanidocarbonat, ethyl cyanoformate, ethyl cyanocarbonate
CCOC(=O)C#N
99.09
99.09
Liquid
Colorless
115°C to 116°C
Authentic
Glass bottle
1.3810 to 1.383
MFCD00001836
02, 547
1
MSMGXWFHBSCQFB-UHFFFAOYSA-N
ethyl cyanoformate
69336
99%
Ethyl cyanoformate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-797-9

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only