missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-4,7,10,13,16,19-Docosahexaenoate 97.0+%, TCI America™

Catalog No. d2195100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2195100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2195100MG Supplier TCI America Supplier No. D2195100MG

Chemical Identifiers

CAS 84494-72-4
Molecular Formula C24H36O2
Molecular Weight (g/mol) 356.55
MDL Number MFCD00056293
InChI Key ITNKVODZACVXDS-UYXIOUCRSA-N
Synonym DHA Ethyl Ester, cis-4,7,10,13,16,19-Docosahexaenoic Acid Ethyl Ester
PubChem CID 71771404
IUPAC Name ethyl (4Z,7E,10E,13E,16E,19E)-docosa-4,7,10,13,16,19-hexaenoate
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)OCC

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 356.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl cis-4,7,10,13,16,19-Docosahexaenoate
TSCA No
Recommended Storage Refrigerator