missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl cis-3-Iodoacrylate (stabilized with Copper chip) 98.0+%, TCI America™
$369.08 - $1069.97
Chemical Identifiers
| CAS | 31930-36-6 |
|---|---|
| Molecular Formula | C5H7IO2 |
| Molecular Weight (g/mol) | 226.01 |
| MDL Number | MFCD00209584,MFCD11519164 |
| InChI Key | AELYFQSZXFFNGP-UHFFFAOYSA-N |
| Synonym | cis-3-Iodoacrylic Acid Ethyl Ester |
| PubChem CID | 4184066 |
| IUPAC Name | ethyl 3-iodoprop-2-enoate |
| SMILES | CCOC(=O)C=CI |
Chemical Identifiers
| 31930-36-6 | |
| 226.01 | |
| AELYFQSZXFFNGP-UHFFFAOYSA-N | |
| 4184066 | |
| CCOC(=O)C=CI |
| C5H7IO2 | |
| MFCD00209584,MFCD11519164 | |
| cis-3-Iodoacrylic Acid Ethyl Ester | |
| ethyl 3-iodoprop-2-enoate |
Specifications
| 31930-36-6 | |
| C5H7IO2 | |
| 5 g | |
| AELYFQSZXFFNGP-UHFFFAOYSA-N | |
| ethyl 3-iodoprop-2-enoate | |
| 4184066 | |
| ≥98.0% (GC) | |
| Ethyl cis-3-Iodoacrylate (stabilized with Copper chip) |
| 85°C | |
| MFCD00209584,MFCD11519164 | |
| cis-3-Iodoacrylic Acid Ethyl Ester | |
| CCOC(=O)C=CI | |
| 226.01 | |
| 226.01 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator