missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-3-Bromoacrylate 98.0+%, TCI America™

Catalog No. B23621G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23621G Supplier TCI America Supplier No. B23621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31930-34-4
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00674501
InChI Key UJTJVQIYRQALIK-ONEGZZNKSA-N
Synonym ethyl cis-3-bromoacrylate, ethyl 2z-3-bromoprop-2-enoate, ethyl z-3-bromoacrylate, ethyl 2z-3-bromoacrylate, ethyl z-3-bromoprop-2-enoate, cis-3-bromoacrylic acid ethyl ester, ethyl cis-3-bromoacrylate gc, 2-propenoic acid, 3-bromo-, ethyl ester, 2z
PubChem CID 643842
IUPAC Name ethyl (2E)-3-bromoprop-2-enoate
SMILES CCOC(=O)\C=C\Br

Specifications

Color Yellow
Boiling Point 67°C
Quantity 1 g
UN Number 3272
Formula Weight 179.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl cis-3-Bromoacrylate
TSCA No
Recommended Storage Refrigerator