missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%

Catalog No. AC298392500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC298392500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC298392500 Supplier Thermo Scientific Chemicals Supplier No. 298392500
Only null left

CAS: 142547-15-7 | C8H16ClNO2 | 193.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142547-15-7
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.67
MDL Number MFCD00800503
InChI Key RYBMXCZWUCHLJB-UHFFFAOYNA-N
Synonym ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, ethyl 1r,2s-2-aminocyclopentane-1-carboxylate hydrochloride, cis-2-amino-cyclopentanecarboxylic acid ethyl ester hydrochloride, rel-1r,2s-ethyl 2-aminocyclopentanecarboxylate hydrochloride, 1r,2s-rel-ethyl 2-aminocyclopentanecarboxylate hydrochloride, ethyl cis-2-aminocyclopentanecarboxylate hcl, cis-ethyl 2-amino-1-cyclopentanecarboxylate hydrochloride, 1r,2s-rel-ethyl2-aminocyclopentanecarboxylatehydrochloride, cis-2-amino-cyclopentanecarboxylic acid ethyl ester hydrochlorid, 1r*,2s*-ethyl 2-aminocyclopentane-1-carboxylate hydrochloride
PubChem CID 2725067
IUPAC Name ethyl (1R,2S)-2-aminocyclopentane-1-carboxylate;hydrochloride
SMILES Cl.CCOC(=O)C1CCCC1N

Specifications

Melting Point 97.0°C to 100.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble in water
Formula Weight 193.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl cis-2-amino-1-cyclopentane carboxylate hydrochloride, 99%
Show More Show Less

RUO – Research Use Only