missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chrysanthemate, 95%, mixture of cis and trans

Catalog No. p-128833
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156060250 25 g Glass bottle
AC156065000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156060250 Supplier Thermo Scientific Chemicals Supplier No. 156060250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-41-6 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-41-6
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00001304
InChI Key VIMXTGUGWLAOFZ-UHFFFAOYSA-N
Synonym ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate
PubChem CID 7334
IUPAC Name ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
SMILES CCOC(=O)C1C(C1(C)C)C=C(C)C

Specifications

Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 112°C (10.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.461
Quantity 25 g
Beilstein 09, II, 45
Specific Gravity 0.9
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl chrysanthemate, 95%
Show More Show Less
EINECSNumber 202-579-7
RTECSNumber GZ1727000
TSCA TSCA

RUO – Research Use Only