missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Chloroglyoxylate 98.0+%, TCI America™

Catalog No. C106625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C106625G 25 g
C1066100G 100 g
C1066500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C106625G Supplier TCI America Supplier No. C106625G

Chemical Identifiers

CAS 4755-77-5
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00000706
InChI Key OWZFULPEVHKEKS-UHFFFAOYSA-N
Synonym ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
PubChem CID 20884
IUPAC Name ethyl 2-chloro-2-oxoacetate
SMILES CCOC(=O)C(=O)Cl

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
UN Number 2920
Formula Weight 136.53
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl Chloroglyoxylate