missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroformate, 99%, Thermo Scientific Chemicals

$220.55 - $220.55

Chemical Identifiers

CAS 541-41-3
MDL Number MFCD00000644
InChI Key RIFGWPKJUGCATF-UHFFFAOYSA-N
Synonym ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
PubChem CID 10928
IUPAC Name ethyl carbonochloridate
SMILES CCOC(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC118255000
SDS View Documents
Thermo Scientific Chemicals
118255000
500 mL Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

541-41-3
RIFGWPKJUGCATF-UHFFFAOYSA-N
10928
CCOC(=O)Cl
MFCD00000644
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
ethyl carbonochloridate

Specifications

541-41-3
1.1350g/mL
10°C
1% max. (diethyl carbonate) (GC)
C3H5ClO2
ClCO2C2H5
500 mL
01,364; 02,193; 04,228; 05,294; 07,147; 09,214; 12,223
15, 3839
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in benzene, chloroform and ether, soluble in most organic solvents
CCOC(=O)Cl
108.52
0.46 mPa.s (20°C)
99%
Ethyl chloroformate, 99%
-81.0°C
94.0°C
Authentic
Glass bottle
1.3950 to 1.3960
MFCD00000644
03, 10
1.135
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
RIFGWPKJUGCATF-UHFFFAOYSA-N
ethyl carbonochloridate
10928
108.52
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

EINECSNumber : 208-778-5

RTECSNumber : LQ6125000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only