missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroformate, 97%

Catalog No. p-7044415
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0631106 5 g
AAL0631122 100 g
AAL0631136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0631106 Supplier Thermo Scientific Chemicals Supplier No. L0631106
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 541-41-3 | C3H5ClO2 | 108.521 g/mol

Ethyl chloroformate is used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction. It is also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles. It is used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroformate is used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction. It is also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles. It is used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.

Solubility
Not miscible in water.

Notes
Protect from heat. Store away from moisture and oxidizing agents. Store under dry inert gas. Refrigerate. Incompatible with heat, moisture and oxidizing agents.

Chemical Identifiers

CAS 541-41-3
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.521
MDL Number MFCD00000644
InChI Key RIFGWPKJUGCATF-UHFFFAOYSA-N
Synonym ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
PubChem CID 10928
IUPAC Name ethyl carbonochloridate
SMILES CCOC(=O)Cl

Specifications

Melting Point -81°C
Density 1.136
Boiling Point 92°C to 93°C
Flash Point 10°C (50°F)
Odor Pungent
Refractive Index 1.395
Linear Formula ClCO2CH2CH3
Quantity 5 g
UN Number UN1182
Beilstein 385653
Sensitivity Moisture sensitive
Merck Index 14,3784
Solubility Information Not miscible in water.
Formula Weight 108.52
Percent Purity 97%
Chemical Name or Material Ethyl chloroformate
Show More Show Less
Hazard Category H225-H301-H314-H330-H335
Hazard Statement GHS H Statement
H225-H330-H314-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYL CHLOROFORMATE
EINECSNumber 208-778-5
RTECSNumber LQ6125000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only