missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Chlorofluoroacetate 98.0+%, TCI America™

Catalog No. C14675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14675G 5 g
C146725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14675G Supplier TCI America Supplier No. C14675G

Chemical Identifiers

CAS 401-56-9
Molecular Formula C4H6ClFO2
Molecular Weight (g/mol) 140.538
MDL Number MFCD00039333
InChI Key WUHVJSONZHSDFC-UHFFFAOYSA-N
Synonym ethyl chlorofluoroacetate, acetic acid, chlorofluoro-, ethyl ester, chlorofluoroacetic acid ethyl ester, ethyl chloro fluoro acetate, ethyl fluorochloroacetate, acetic acid, 2-chloro-2-fluoro-, ethyl ester, acmc-1csjg, ksc492m6l, ethyl 2-chloro-2-fluoro-acetate
PubChem CID 101668
IUPAC Name ethyl 2-chloro-2-fluoroacetate
SMILES CCOC(=O)C(F)Cl

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
UN Number 2924
Formula Weight 140.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Chlorofluoroacetate