missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Chloroacetate 98.0+%, TCI America™

Catalog No. p-7124213
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C009225ML 25 mL
C0092500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C009225ML Supplier TCI America Supplier No. C009225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-39-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00000932
InChI Key VEUUMBGHMNQHGO-UHFFFAOYSA-N
Synonym ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf
PubChem CID 7751
IUPAC Name ethyl 2-chloroacetate
SMILES CCOC(=O)CCl

Specifications

Melting Point -26°C
Color Colorless
Boiling Point 143°C
Quantity 25 mL
UN Number 1181
Formula Weight 122.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Chloroacetate
EINECSNumber (2)-1149
RTECSNumber AF9110000
TSCA Yes