missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroacetate, 99%

Catalog No. p-3736427
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118222500 250 mL Glass bottle
AC118220010 1 L Glass bottle
AC118220025 2.5 L Glass bottle
AC118220100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC118222500 Supplier Thermo Scientific Chemicals Supplier No. 118222500
Only null left

CAS: 105-39-5 | C4H7ClO2 | 122.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-39-5
MDL Number MFCD00000932
InChI Key VEUUMBGHMNQHGO-UHFFFAOYSA-N
Synonym ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf
PubChem CID 7751
IUPAC Name ethyl 2-chloroacetate
SMILES CCOC(=O)CCl

Specifications

Melting Point -26.0°C
Color Colorless
Density 1.1500g/mL
Boiling Point 142.0°C to 145.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7ClO2
Refractive Index 1.4200 to 1.4220
Linear Formula ClCH2CO2C2H5
Quantity 250 mL
Beilstein 02,197
Fieser 13,348
Specific Gravity 1.15
Merck Index 15,3838
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
Molecular Weight (g/mol) 122.55
Viscosity 1.27 mPa.s (20°C)
Formula Weight 122.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl chloroacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at
EINECSNumber 203-294-

RUO – Research Use Only