missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethyl cellulose, ethoxyl content 48%, 22 cps

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol

Supplier:  Thermo Scientific Chemicals 190972500

 View more versions of this product

Catalog No. AC190972500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl cellulose
9004-57-3
MFCD00131037
Solubility in water: insoluble. Other solubilities: inert to alkalies and dilute acids, soluble in most organic liquids, insoluble in glycerol
ARSKJXYLLONUAJ-UHFFFAOYSA-N
2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
166304
1.47
Ethoxyl Content 48%, 22 Cps
C23H24N6O4
n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
250 g
CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
448.483
Glass bottle

Chemical Identifiers

9004-57-3
448.483
ARSKJXYLLONUAJ-UHFFFAOYSA-N
166304
CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
C23H24N6O4
MFCD00131037
n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : FJ5950500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only