missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Ethyl cellulose, ethoxyl content 48%, 22 cps
Supplier: Thermo Scientific™ 190970025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Ethyl cellulose | |
| 9004-57-3 | |
| MFCD00131037 | |
| 2.5kg | |
| CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C | |
| 448.483 | |
| Plastic drum |
| Ethoxyl Content 48%, 22 Cps | |
| C23H24N6O4 | |
| n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene | |
| ARSKJXYLLONUAJ-UHFFFAOYSA-N | |
| 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate | |
| 166304 | |
| 1.47 |
Safety and Handling
RTECSNumber : FJ5950500
TSCA : TSCA
RUO â Research Use Only