missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethyl cellulose

Catalog No. aah27621md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27621MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH27621MD Supplier Thermo Scientific Chemicals Supplier No. H27621MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol

Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color. In addition to this, it is used in lacquers and plastic toughing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color.

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.483
MDL Number MFCD00131037
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
Synonym n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
PubChem CID 166304
IUPAC Name 2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
SMILES CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C

Specifications

Density 1.14
Refractive Index 1.47
Quantity 250 mg
Solubility Information Soluble in tetrahydrofuran,methyl acetate,ethanol,methanol,toluene,ethyl acetate,chloroform,esters,aromatic hydrocarbons,alcohols and ketones. Insoluble in water,glycerol and propane-1,2-diol.
Chemical Name or Material Ethyl cellulose
RTECSNumber FJ5950500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only