missing translation for 'onlineSavingsMsg'
Learn More

Ethyl carbazate, 97%

CAS: 4114-31-2 | C3H8N2O2 | 104.109 g/mol

Supplier:  Thermo Scientific Chemicals A138600B

 View more versions of this product

Catalog No. AAA138600B

Only null left
Explore available promotions

Description

Description

Prepare 4-phenylurazole from ethyl carbazate and then oxidize the urazole with gaseous dinitrogen tetroxide to yield 4-phenyl-1,2,4-tri- azoline-3,5-dione la. Modification of the ethyl carbazate procedure in which analytically pure monoalkylhydrazine hydrochlorides were isolated in greater than 90% yield, starting from a ketone or aldehyde. Treatment of primary amines is with chloroamine or hydroxylamine-0-sulfonic acid and the condensation of a carbonyl compound with ethyl carbazate. The FeCl 3 -catalyzed reaction of ethyl carbazate (2 b) proceeded readily to give β-hydroxyester 3 b in better yield. A new one-pot neat synthesis of some 5-aryl-2,4-dihydro-3H-1,2,4-triazol-3-ones through cyclocondensation of ethyl carbazate with aryl nitriles catalyzed by DMAP as an efficient and basic nucleophilic catalyst is described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Prepare 4-phenylurazole from ethyl carbazate and then oxidize the urazole with gaseous dinitrogen tetroxide to yield 4-phenyl-1,2,4-tri- azoline-3,5-dione la. Modification of the ethyl carbazate procedure in which analytically pure monoalkylhydrazine hydrochlorides were isolated in greater than 90% yield, starting from a ketone or aldehyde. Treatment of primary amines is with chloroamine or hydroxylamine-0-sulfonic acid and the condensation of a carbonyl compound with ethyl carbazate. The FeCl 3 -catalyzed reaction of ethyl carbazate (2 b) proceeded readily to give β-hydroxyester 3 b in better yield. A new one-pot neat synthesis of some 5-aryl-2,4-dihydro-3H-1,2,4-triazol-3-ones through cyclocondensation of ethyl carbazate with aryl nitriles catalyzed by DMAP as an efficient and basic nucleophilic catalyst is described.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Ethyl carbazate
44°C to 47°C
86°C (186°F)
MFCD00007595
UN2811
Moisture sensitive
Very soluble in water.
CCOC(=O)NN
104.109
104.11
4114-31-2
85°C to 86°C (7 mmHg)
C3H8N2O2
1000 g
878265
ethyl carbazate, ethyl hydrazinecarboxylate, ethoxycarbohydrazide, hydrazinecarboxylic acid, ethyl ester, carbethoxyhydrazine, ethylcarbazate, ethyl carbazinate, carboethoxyhydrazine, monocarbethoxyhydrazine, n-carbethoxy hydrazine
VYSYZMNJHYOXGN-UHFFFAOYSA-N
ethyl N-aminocarbamate
20064
97%

Chemical Identifiers

4114-31-2
104.109
VYSYZMNJHYOXGN-UHFFFAOYSA-N
20064
CCOC(=O)NN
C3H8N2O2
MFCD00007595
ethyl carbazate, ethyl hydrazinecarboxylate, ethoxycarbohydrazide, hydrazinecarboxylic acid, ethyl ester, carbethoxyhydrazine, ethylcarbazate, ethyl carbazinate, carboethoxyhydrazine, monocarbethoxyhydrazine, n-carbethoxy hydrazine
ethyl N-aminocarbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 223-903-3

RTECSNumber : FE2545000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only