missing translation for 'onlineSavingsMsg'
Learn More

Ethyl butyrylacetate, 98%

Catalog No. p-7020729
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1278618 50 g
AAA1278630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1278618 Supplier Thermo Scientific Chemicals Supplier No. A1278618
Only null left

CAS: 3249-68-1 | C8H14O3 | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3249-68-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009401
InChI Key KQWWVLVLVYYYDT-UHFFFAOYSA-N
Synonym ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683
PubChem CID 238498
ChEBI CHEBI:18119
IUPAC Name ethyl 3-oxohexanoate
SMILES CCCC(=O)CC(=O)OCC

Specifications

Melting Point -44°C
Density 0.99
Boiling Point 94°C to 95°C (12 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.427
Quantity 50 g
Beilstein 507689
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material Ethyl butyrylacetate
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 221-835-9
RTECSNumber MO8420500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only