missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromodifluoroacetate, 98%

Catalog No. p-127448
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305800050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC305800050 Supplier Thermo Scientific Chemicals Supplier No. 305800050

CAS: 667-27-6 | C4H5BrF2O2 | 202.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 667-27-6
Molecular Formula C4H5BrF2O2
Molecular Weight (g/mol) 202.99
MDL Number MFCD00042069
InChI Key IRSJDVYTJUCXRV-UHFFFAOYSA-N
Synonym ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953
PubChem CID 69585
IUPAC Name ethyl 2-bromo-2,2-difluoroacetate
SMILES CCOC(=O)C(F)(F)Br

Specifications

Color Colorless to Yellow
Density 1.5830g/mL
Boiling Point 112.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3860 to 1.3890
Linear Formula BrCF2CO2C2H5
Quantity 5 g
Specific Gravity 1.583
Solubility Information Solubility in water: insoluble
Formula Weight 202.99
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl bromodifluoroacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 211-567-0

RUO – Research Use Only