missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzoylformate, 95%, Thermo Scientific™

Catalog No. p-3212685
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410350050 5g Glass bottle
AC410350250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC410350050 Supplier Thermo Scientific Chemicals Supplier No. 410350050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1603-79-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00009120
InChI Key QKLCQKPAECHXCQ-UHFFFAOYSA-N
Synonym ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
PubChem CID 15349
ChEBI CHEBI:84260
IUPAC Name ethyl 2-oxo-2-phenylacetate
SMILES CCOC(=O)C(=O)C1=CC=CC=C1

Specifications

Density 1.1220g/mL
Boiling Point 138°C to 139°C (18.0mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5150 to 1.517
Linear Formula C6H5COCO2CH2CH3
Quantity 5g
Specific Gravity 1.122
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material Ethyl benzoylformate
Show More Show Less
EINECSNumber 216-504-

RUO – Research Use Only