missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Benzoylacetate 95.0+%, TCI America™
Supplier: TCI America B0097100G
Specifications
| Ethyl Benzoylacetate | |
| Yellow | |
| C11H12O3 | |
| 100 g | |
| GKKZMYDNDDMXSE-UHFFFAOYSA-N | |
| ethyl 3-oxo-3-phenylpropanoate | |
| 7170 | |
| ≥95.0% (GC,T) |
| 94-02-0 | |
| 270°C | |
| MFCD00009196 | |
| ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate | |
| CCOC(=O)CC(=O)C1=CC=CC=C1 | |
| 192.214 | |
| 192.21 | |
| Liquid |
Chemical Identifiers
| 94-02-0 | |
| 192.214 | |
| GKKZMYDNDDMXSE-UHFFFAOYSA-N | |
| 7170 | |
| CCOC(=O)CC(=O)C1=CC=CC=C1 |
| C11H12O3 | |
| MFCD00009196 | |
| ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate | |
| ethyl 3-oxo-3-phenylpropanoate |
Safety and Handling
EINECSNumber : (3)-1282&(3)-1708&(3)-1743
RTECSNumber : AF4878000
TSCA : Yes