missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzoylacetate 95.0+%, TCI America™

Catalog No. B0097500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0097500G 500 g
B009725G 25 g
B0097100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0097500G Supplier TCI America Supplier No. B0097500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-02-0
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00009196
InChI Key GKKZMYDNDDMXSE-UHFFFAOYSA-N
Synonym ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate
PubChem CID 7170
IUPAC Name ethyl 3-oxo-3-phenylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 270°C
Quantity 500 g
Formula Weight 192.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl Benzoylacetate
EINECSNumber (3)-1282&(3)-1708&(3)-1743
RTECSNumber AF4878000
TSCA Yes