missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzothiazole-2-carboxylate, 98%

Catalog No. p-7031728
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3290406 5 g
AAH3290414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3290406 Supplier Thermo Scientific Chemicals Supplier No. H3290406
Only null left

CAS: 32137-76-1 | C10H9NO2S | 207.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32137-76-1
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.247
MDL Number MFCD00848360
InChI Key VLQLCEXNNGQELL-UHFFFAOYSA-N
Synonym ethyl benzo d thiazole-2-carboxylate, ethyl benzothiazole-2-carboxylate, benzothiazole-2-carboxylic acid ethyl ester, 2-benzothiazolecarboxylic acid, ethyl ester, ethyl 2-benzothiazolecarboxylate, ethyl1,3-benzothiazole-2-carboxylate, zlchem 880, pubchem23783, acmc-209hra, ksc490c5n
PubChem CID 640708
IUPAC Name ethyl 1,3-benzothiazole-2-carboxylate
SMILES CCOC(=O)C1=NC2=CC=CC=C2S1

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
Formula Weight 207.25
Percent Purity 98%
Chemical Name or Material Ethyl benzothiazole-2-carboxylate
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only