missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzimidazole-4-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH66669MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66669MD 250 mg
AAH6666903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66669MD Supplier Thermo Scientific Chemicals Supplier No. H66669MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167487-83-4
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD18251681,MFCD20692524
InChI Key BIROFVBTDWNLKU-UHFFFAOYSA-N
Synonym ethyl 1h-benzo d imidazole-7-carboxylate, ethyl 4-benzimidazolecarboxylate, ethyl 3h-benzo d imidazole-4-carboxylate, ethyl 1h-benzo d imidazole-4-carboxylate, 1h-benzimidazole-4-carboxylic acid, ethyl ester, 1h-benzimidazole-7-carboxylic acid, ethyl ester, ethyl 1h-1,3-benzodiazole-4-carboxylate, ethyl4-benzimidazolecarboxylate, 1h-benzimidazole-4-carboxylicacid,ethylester, 1h-benzimidazole-4-carboxylic acid ethyl ester
PubChem CID 13180239
IUPAC Name ethyl 1H-1,3-benzodiazole-4-carboxylate
SMILES CCOC(=O)C1=C2N=CNC2=CC=C1

Specifications

Quantity 250 mg
Beilstein 9390984
Formula Weight 190.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl benzimidazole-4-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only