missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Benzenesulfonate 98.0+%, TCI America™
$85.75 - $844.41
Chemical Identifiers
| CAS | 515-46-8 |
|---|---|
| Molecular Formula | C8H10O3S |
| Molecular Weight (g/mol) | 186.225 |
| MDL Number | MFCD00025033 |
| InChI Key | XDRMBCMMABGNMM-UHFFFAOYSA-N |
| Synonym | Benzenesulfonic Acid Ethyl Ester |
| PubChem CID | 10585 |
| IUPAC Name | ethyl benzenesulfonate |
| SMILES | CCOS(=O)(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 515-46-8 | |
| 186.225 | |
| XDRMBCMMABGNMM-UHFFFAOYSA-N | |
| 10585 | |
| CCOS(=O)(=O)C1=CC=CC=C1 |
| C8H10O3S | |
| MFCD00025033 | |
| Benzenesulfonic Acid Ethyl Ester | |
| ethyl benzenesulfonate |
Specifications
| 515-46-8 | |
| 156°C | |
| MFCD00025033 | |
| Benzenesulfonic Acid Ethyl Ester | |
| CCOS(=O)(=O)C1=CC=CC=C1 | |
| 186.225 | |
| 186.23 | |
| Liquid |
| Yellow | |
| C8H10O3S | |
| 25 g | |
| XDRMBCMMABGNMM-UHFFFAOYSA-N | |
| ethyl benzenesulfonate | |
| 10585 | |
| ≥98.0% (GC) | |
| Ethyl Benzenesulfonate |
Safety and Handling
TSCA : No