missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha-bromophenylacetate, 97%

Catalog No. p-4229206
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438060050 5 g Glass bottle
AC438060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438060050 Supplier Thermo Scientific Chemicals Supplier No. 438060050

CAS: 2882-19-1 | C10H11BrO2 | 243.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2882-19-1
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.10
MDL Number MFCD00013536
InChI Key BKTKLDMYHTUESO-UHFFFAOYNA-N
Synonym ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate
PubChem CID 97780
IUPAC Name ethyl 2-bromo-2-phenylacetate
SMILES CCOC(=O)C(Br)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.3890g/mL
Boiling Point 89°C to 92°C (1.2 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5385 to 1.5405 (20°C, 589nm)
Linear Formula C6H5Ch(Br)COOCH2CH3
Quantity 5 g
Specific Gravity 1.389
Formula Weight 243.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl α-bromophenylacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-735-2

RUO – Research Use Only